missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress X-GAL

Product Code. 16419467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419467 Supplier MedChemExpress Supplier No. HY15934

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress X-GAL is a widely used chromogenic β-galactosidase substrate. β-galactosidase cleaves X-gal and produce an insoluble blue compound, which is detectable.

Specifications

CAS 7240-90-6
Solubility DMSO : 100 mg/mL (244.72 mM∣Need ultrasonic)
Synonym BCIG
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Quantity 5 g
Molecular Weight (g/mol) 408.63 g/mol
Product Type Dye Reagents
Purity or Quality Grade 95.66%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.