missing translation for 'onlineSavingsMsg'
Learn More

Melford X-GAL

Product Code. p-200087030
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
10 g
100 g
Unit Size:
100g
100mg
10g
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 30018093

Brand: Melford B7180010.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromogenic substrate of β-Galactosidase used in combination with IPTG for detection of β-Galactosidase activity in bacterial colonies.

Chromogenic substrate of β-Galactosidase used in combination with IPTG for detection of β-Galactosidase activity in bacterial colonies. X-GAL is cleaved at the B1-4 bond between galactose and the 5-Bromo-4-chloro-3-indolyl portion of X-GAL by β-Galactosidase via hydrolysis. The cleavage of X-GAL results in the production of a water insoluble blue dichloro-dibromo-indigo precipitate. In cloning strategies with vectors such as Lambda-11, M13mp18 and 19, pUC18 and 19, pUR222 the E. coli lacZ gene is transformed to lac+ cells. After transformation the cells show β-Galactosidase activity in the presence of media containing X-GAL and IPTG. The insertion of a DNA fragment into the cloning sites of the lacZ gene results in the disruption of β-Galactosidase activity leading to the appearance of white colonies. Non-recombinant cells produce a blue indigo dye.

Water Content: 0.05% (K. Fisher)

Protect from light and moisture

Specifications

Quantity 10 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.