missing translation for 'onlineSavingsMsg'
Learn More

Klever X-Change (Wider Cut Head)

Product Code. 17709007
Click to view available options
Color:
Blue
Orange
Red
Yellow
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 17709007

Brand: Klever KCJXC30R

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Recessed blade of the Klever X-Change (Wider Cut Head) reduces cut injuries and damaged goods.

  • Cuts boxes, film, tape and thousands more applications.
  • Ergonomic 'soft touch' handle reduces hand fatigue and stress.
  • Innovative tape splitter.
  • Adavnced plastic polymers.
  • Highest safety level.
  • Interchangeable, replaceable head.

Specifications

Product Type Klever X Change Wide Head
For Use With (Application) Cuts Boxes, Film, Tape, and Thousands More Applications
Disposable Reusable
Sterility Non-sterile
Color Red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.