missing translation for 'onlineSavingsMsg'
Learn More

Draeger X-am™ 7000 Portable Multi Gas Detector

Product Code. 16401255
Change view
Click to view available options
Components Measured:
Oxygen, Toxic and Combustible Gases and Vapors
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured
16401255 Oxygen, Toxic and Combustible Gases and Vapors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401255 Supplier Draeger Supplier No. 3310839

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the simultaneous and continuous measurement of up to five gases. It is the ideal companion in a variety of applications where the reliable detection of oxygen, toxic and combustible gases and vapors are necessary.

  • Flexibility through sensor variety
  • Intelligent and interchangeable sensors
  • Poison resistant ex-sensor
  • Robust and watertight
  • Built for longevity
  • Multiple alarms
  • Strong integrated pump
  • Easy detection of leakages
  • Information at a glance
  • Intuitive software
  • Data management

Specifications

Alarm Types Visual 360° flashing LEDs, audible alarm (>100 dB (at 30 cm or 1 ft.))
Battery Life NiMH (4.8 V / 3.0 Ah) >9 hrs, NiMH (4.8 V / 6.0 Zh) > 20 hrs, Alkaline > 20 hrsCharging time: 3.5 to 7 hours, depending on battery type
Components Measured Oxygen, Toxic and Combustible Gases and Vapors
Dimensions (L x W x H) 150 x 140 x 75 mm
For Use With (Application) Detection of Oxygen, Toxic and Combustible Gases and Vapours
Humidity Range 10 to 95 % r.h.
Includes Calibration adapter, water and dust filter. Requires (but not included) power Pk. and up to 5 sensors. Optional integrated pump or data logger can also be included, please enqure.
Input Pressure (Zero and 5 Span Gases) 700 to 1300 hPa
IP Rating IP67
Temperature Range (Metric) -20°C to +55°C, short-term -40°C to +60°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.