missing translation for 'onlineSavingsMsg'
Learn More

Melford X-a-Gal

Product Code. p-200087002
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 30018097

Brand: Melford B719001.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromogenic substrate for detection of a-galactosidase activity in yeast Saccharomyces cerevisiae.

Chromogenic substrate for detection of a-galactosidase activity in yeast Saccharomyces cerevisiae. Used in yeast Two-Hybrid System to detect protein-protein interactions within random libraries. The enzyme a-galactosidase enabels yeast to use disaccharide melibose as a carbon source during growth or fermentation. Secretion of this enzyme in response to activation of the GAL4 gene hydrolizes X-a-GAL and causes yeast colonies to develop a blue color. This reporter system allows immediate visible results on selection plates and eliminates necessity of a second screen filter life assay.

Specifications

Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.