missing translation for 'onlineSavingsMsg'
Learn More

WZ4141R, MedChemExpress

Product Code. 16450989
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450989 1 mg 1mg
16446497 5 mg 5mg
16424836 100 mg 100mg
16410157 50 mg 50mg
16403778 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16450989 Supplier Medchem Express Supplier No. HY103016
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress WZ4141R is an intermediate in the synthesis of compounds.

WZ4141R is an intermediate in the synthesis of compounds.

Specifications

Chemical Name or Material WZ4141R
CAS 2319589-98-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H15N3O2
Quantity 1 mg
SMILES O=C(CC)NC1=CC(OC2=C3C(NC=C3)=NC=C2)=CC=C1
Molecular Weight (g/mol) 281.31
Formula Weight 281.31
Percent Purity 98.03%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.