missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ WypAll™ L10 Light Duty Cleaning Paper Wipes

Product Code. 31188848
Change view
Click to view available options
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
31188848 Blue
31188851 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Plastic packaging - suffocation hazard. Please ensure to carry the product in a safe manner for the weight indicated on the packaging. For food contact.
Product Code. 31188848 Supplier Kimberly-Clark Professional™ Supplier No. 7378
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Plastic packaging - suffocation hazard. Please ensure to carry the product in a safe manner for the weight indicated on the packaging. For food contact.

WypAll™ L10 compact wiping paper is designed for light-duty hygiene wiping and perfect for tasks like cleaning food preparation and service surfaces. The embossing technology of these paper wipes delivers a great balance of performance and value.

No matter which surfaces they come into contact with throughout their day, give your employees and visitors greater peace of mind by providing proven cleaning and disinfecting workplace hygiene solutions to help break the chain of germ transmission. WypAll™ L10 professional wiping paper products are designed for the specific environments where they will be used, helping enable you to deliver on your wiping task every time. These compact disposable paper wipes are 1 ply and designed for light duty wiping in areas such as food preparation and service surfaces. The compact design of this wiping paper is designed to be small, portable and easy to handle. This formats ease of use helps to support hygiene standards and compliance while also saving space. Embossing technology in the wiping paper gives these cleaning wipes a great balance of performance and value. This WypAll™ wiping paper is designed to be used once with detergent or disinfectant. These dry cleaning wipes have been tested as compatible for use in spray and wipe application with peracetic acid, hydrogen peroxide and quats (quaternary ammonium compounds). Spray and wiping compatibility helps to ensure workers can wipe own work spaces, vehicles, desks, seating and food prep areas before and after use. Spray and wipe also helps to enable production workers to clean down/sanitise individual workstations before hand-off to next shift. For environments where fully saturated wiping is a requirement, these WypAll™ wiping rolls are tested as compatible with soap/detergent. Always follow manufacturers instructions for dilution, application and contact time.

  • Individual sheet size 25 (L) x 34 cm (W)
  • Each WypAll™ L10 compact wiping roll contains 165 wiping paper sheets
  • Dry cleaning wipes designed for light duty hygiene wiping and perfect for tasks like cleaning food preparation and service surfaces
  • Embossing technology in the wiping paper gives these cleaning wipes a great balance of performance and value
  • These WypAll™ paper wipers come in a compact roll format for portability and ease of use
  • Designed for use with the WypAll™ wiping paper dispenser (product code 7041)
  • Help break the chain of germ transmission
TRUSTED_SUSTAINABILITY

Specifications

Product Type Wiping Paper
Length (English) 9.84 in.
Length (Metric) 25 cm
Width (English) 13.38 in.
Width (Metric) 34 cm
Material Paper
Color Blue
Dimensions (L x W) 25 x 34 cm
Packaging Quantity 12 Rolls x 165 Sheets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.