missing translation for 'onlineSavingsMsg'
Learn More

WYE-354, MedChemExpress

Product Code. 16457677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457677 5 mg 5mg
16434938 10 mg 10mg
16401187 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16457677 Supplier Medchem Express Supplier No. HY120345mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WYE-354 is an ATP-competitive mTOR inhibitor with an IC50 of 5 nM. WYE-354 also inhibits PI3Kα and PI3Kγ with IC50s of 1.89 μM and 7.37 μM, respectively. WYE-354 inhibits both mTORC1 and mTORC2. WYE-354 induces autophagy activation in vitro.

WYE-354 is an ATP-competitive mTOR inhibitor with an IC50 of 5 nM. WYE-354 also inhibits PI3Kα and PI3Kγ with IC50s of 1.89 μM and 7.37 μM, respectively. WYE-354 inhibits both mTORC1 and mTORC2. WYE-354 induces autophagy activation in vitro.

Specifications

Chemical Name or Material WYE-354
CAS 1062169-56-5
Color White
Purity Grade Notes Research
Molecular Formula C24H29N7O5
Quantity 5 mg
Solubility Information DMSO : 6.67 mg/mL (13.46 mM; Need ultrasonic)
SMILES COC(NC1=CC=C(C=C1)C2=NC3=C(C(N4CCOCC4)=N2)C=NN3C5CCN(CC5)C(OC)=O)=O
Molecular Weight (g/mol) 495.53
Formula Weight 495.53
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.