missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WY 14643

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective PPARα agonist (EC50 values are 0.63, 32 and > 100 μM at PPARα, PPARγ and PPARδ respectively). Negatively inhibits NF-κB transcriptional activity and decreases the inflammatory response in vitro and in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 323.8 |
| Purity | >98% |
| Molecular Weight (g/mol) | 323.80 |
| Chemical Name or Material | WY 14643 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto