missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95211-0.02ml
This item is not returnable.
View return policy
Description
WWP2 Polyclonal antibody specifically detects WWP2 in Human, Rat samples. It is validated for Western Blot
Specifications
| WWP2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| AIP2Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 2, Atrophin-1-interacting protein 2, EC 6.3.2, EC 6.3.2.-, NEDD4-like E3 ubiquitin-protein ligase WWP2, WW domain containing E3 ubiquitin protein ligase 2, WW domain-containing protein 2, WWp2-like | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human WWP2 (NP_001257383.1). MASASSSRAGVALPFEKSQLTLKVVSAKPKVHNRQPRINSYVEVAVDGLPSETKKTGKRIGSSELLWNEIIILNVTAQSHLDLKVWSCHTLRNELLGTASVNLSNVLKNNGGKMENMQLTLNLQTENKGSVVSGGELTIFLDGPTVDLGNVPNGSALTDGSQLPSRDSSGTAVAPENRHQPPSTNCFGGRSRTHRHSGASARTTPATGEQSPGARSRHRQPVKNSGHSGLANGTVNDEPTTATDPEEPSVVGVTSPPAAPLSVTPNPNTT | |
| 0.02 mL | |
| Cell Biology | |
| 11060 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction