missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWP2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49942AF750
This item is not returnable.
View return policy
Description
WWP2 Polyclonal antibody specifically detects WWP2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| WWP2 | |
| Polyclonal | |
| Western Blot | |
| AIP2Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 2, Atrophin-1-interacting protein 2, EC 6.3.2, EC 6.3.2.-, NEDD4-like E3 ubiquitin-protein ligase WWP2, WW domain containing E3 ubiquitin protein ligase 2, WW domain-containing protein 2, WWp2-like | |
| The immunogen for this product maps to a region between residue 200 and 250 of human WW Domain Containing E3 Ubiquitin Protein Ligase 2 using the numbering given in entry NP_008945.2 (GeneID 11060). | |
| 0.1 mL | |
| Cell Biology | |
| 11060 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction