missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49713
This item is not returnable.
View return policy
Description
WWP1 Polyclonal specifically detects WWP1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| WWP1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| AIP5Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 5, Atrophin-1-interacting protein 5, DKFZP434D2111, EC 6.3.2, EC 6.3.2.-, hSDRP1, NEDD4-like E3 ubiquitin-protein ligase WWP1, TGIF-interacting ubiquitin ligase 1, TIUL1, WW domain containing E3 ubiquitin protein ligase 1, WW domain-containing protein 1 | |
| Rabbit | |
| 111 kDa | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative) | |
| Q9H0M0 | |
| WWP1 | |
| The immunogen for this product maps to a region between residue 300 and 350 of human WW Domain Containing E3 Ubiquitin Protein Ligase 1 using the numbering given in entry NP_008944.1 (GeneID 11059). | |
| Affinity Purified | |
| RUO | |
| 11059 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction