missing translation for 'onlineSavingsMsg'
Learn More

WWP1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18404434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404434 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404434 Supplier Novus Biologicals Supplier No. NBP149713AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WWP1 Polyclonal antibody specifically detects WWP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen WWP1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AIP5Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 5, Atrophin-1-interacting protein 5, DKFZP434D2111, EC 6.3.2, EC 6.3.2.-, hSDRP1, NEDD4-like E3 ubiquitin-protein ligase WWP1, TGIF-interacting ubiquitin ligase 1, TIUL1, WW domain containing E3 ubiquitin protein ligase 1, WW domain-containing protein 1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 300 and 350 of human WW Domain Containing E3 Ubiquitin Protein Ligase 1 using the numbering given in entry NP_008944.1 (GeneID 11059).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11059
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.