missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WWOX Recombinant Rabbit Monoclonal Antibody (K01_2Q01)
GREENER_CHOICE

Product Code. 30829288
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30829288 100 μL 100µL
30828144 50 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829288 Supplier Invitrogen™ Supplier No. MA564238

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

WWOX Recombinant Monoclonal Antibody for IHC (P), Western Blot

WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 2 WW domains and a short-chain dehydrogenase/reductase domain (SRD). The highest normal expression of this gene is detected in hormonally regulated tissues such as testis, ovary, and prostate. This expression pattern and the presence of an SRD domain suggest a role for this gene in steroid metabolism. The encoded protein is more than 90% identical to the mouse protein, which is an essential mediator of tumor necrosis factor-alpha-induced apoptosis, suggesting a similar, important role in apoptosis for the human protein. In addition, there is evidence that this gene behaves as a suppressor of tumor growth. Alternative splicing of this gene generates transcript variants that encode different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WWOX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2Q01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Wwox
Gene Accession No. Q91WL8, Q9NZC7
Gene Alias 5330426P09Rik; 9030416C10Rik; D16S432E; EIEE28; FOR; FRA16D; Fragile site FRA16D oxidoreductase; HHCMA56; PRO0128; SCAR12; SDR41C1; Short chain dehydrogenase/reductase family 41C member 1; short chain dehydrogenase/reductase family 41C, member 1; WOX1; WW domain containing oxidoreductase; WW domain-containing oxidoreductase; WW domain-containing protein WWOX; WW-domain oxidoreductase; Wwox
Gene Symbols Wwox
Host Species Rabbit
Immunogen Recombinant protein of human WWOX.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51741, 80707
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.