missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WWOX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529701
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, Raji, mouse brain, rat brain, HepG2. Predicted reactivity: Human (99%), Mouse (95%), Rat (96%), Chicken (83%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 2 WW domains and a short-chain dehydrogenase/reductase domain (SRD). The highest normal expression of this gene is detected in hormonally regulated tissues such as testis, ovary, and prostate. This expression pattern and the presence of an SRD domain suggest a role for this gene in steroid metabolism. The encoded protein is more than 90% identical to the mouse protein, which is an essential mediator of tumor necrosis factor-alpha-induced apoptosis, suggesting a similar, important role in apoptosis for the human protein. In addition, there is evidence that this gene behaves as a suppressor of tumor growth. Alternative splicing of this gene generates transcript variants that encode different isoforms.
Specifications
| WWOX | |
| Polyclonal | |
| Unconjugated | |
| Wwox | |
| 5330426P09Rik; 9030416C10Rik; D16S432E; EIEE28; FOR; FRA16D; Fragile site FRA16D oxidoreductase; HHCMA56; PRO0128; SCAR12; SDR41C1; Short chain dehydrogenase/reductase family 41C member 1; short chain dehydrogenase/reductase family 41C, member 1; WOX1; WW domain containing oxidoreductase; WW domain-containing oxidoreductase; WW domain-containing protein WWOX; WW-domain oxidoreductase; Wwox | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 292041, 51741, 80707 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q91WL8, Q9NZC7 | |
| Wwox | |
| Recombinant protein encompassing a sequence within the center region of human WWOX. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction