missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWOX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32122
This item is not returnable.
View return policy
Description
WWOX Polyclonal specifically detects WWOX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| WWOX | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| D16S432E, EC 1.1.1, EC 1.1.1.-, FORHHCMA56, FRA16D, Fragile site FRA16D oxidoreductase, SDR41C1, short chain dehydrogenase/reductase family 41C, member 1, WOX1PRO0128, WW domain containing oxidoreductase, WW domain-containing oxidoreductase, WW domain-containing protein WWOX | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunoprecipitation 2 - 10 mcg/mg lysate | |
| P_057457.1 | |
| WWOX | |
| The immunogen this antibody was made to, maps to a region between residue 364 to 414 of human WW Domain Containing Oxidoreductase using the numbering given in entry P_057457.1 (GeneID 51741). | |
| 100 μL | |
| Alzheimers Research, Apoptosis, Cancer, Neuroscience, Tumor Suppressors | |
| 51741 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction