missing translation for 'onlineSavingsMsg'
Learn More

WWOX Antibody, DyLight 405, Novus Biologicals™

Product Code. 18416637 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416637 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416637 Supplier Novus Biologicals Supplier No. NBP232122V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WWOX Polyclonal antibody specifically detects WWOX in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen WWOX
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias D16S432E, EC 1.1.1, EC 1.1.1.-, FORHHCMA56, FRA16D, Fragile site FRA16D oxidoreductase, SDR41C1, short chain dehydrogenase/reductase family 41C, member 1, WOX1PRO0128, WW domain containing oxidoreductase, WW domain-containing oxidoreductase, WW domain-containing protein WWOX
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 364 to 414 of human WW Domain Containing Oxidoreductase using the numbering given in entry P_057457.1 (GeneID 51741).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51741
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.