missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWOX Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32121AF405
This item is not returnable.
View return policy
Description
WWOX Polyclonal antibody specifically detects WWOX in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| WWOX | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| D16S432E, EC 1.1.1, EC 1.1.1.-, FORHHCMA56, FRA16D, Fragile site FRA16D oxidoreductase, SDR41C1, short chain dehydrogenase/reductase family 41C, member 1, WOX1PRO0128, WW domain containing oxidoreductase, WW domain-containing oxidoreductase, WW domain-containing protein WWOX | |
| The immunogen this antibody was made to, maps to a region between residue 75 to 125 of human WW Domain Containing Oxidoreductase using the numbering given in entry P_057457.1 (GeneID 51741). | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cancer, Neuroscience, Tumor Suppressors | |
| 51741 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction