missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WWC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560582
This item is not returnable.
View return policy
Description
Immunogen sequence: LRVDLCSVSK HRREECLAGT QISLADLPFS SEVFTLWYNL LPSKQMPCKK NEENEDSVFQ PNQPLVDSID LDAVSALLAR TSAELLAVEQ ELAQEEEEES G Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.
This gene encodes a member of the WW-and-C2-domain-containing family of proteins. Members of this family have two N-terminal WW domains that mediate binding to target proteins harboring L/PPxY motifs, an internal C2 domain for membrane association, and C-terminal alpha protein kinase C binding sites and class III PDZ domain-interaction motifs. Proteins of this family are able to form homo- and heterodimers and to modulate hippo pathway signaling.
Specifications
| WWC2 | |
| Polyclonal | |
| Unconjugated | |
| WWC2 | |
| 5330431C14Rik; AU022508; BH-3-only member B; BH3-only member B protein; BOMB; C85457; D8Ertd594e; LOW QUALITY PROTEIN: protein WWC2; MNCb-4173; Protein WWC2; RGD1559427; WW and C2 domain containing 2; WW domain-containing protein 2; WW, C2 and coiled-coil domain containing 2; WWC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80014 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6AWC2 | |
| WWC2 | |
| Recombinant protein corresponding to Human WWC2. Recombinant protein control fragment (Product #RP-100156). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido