missing translation for 'onlineSavingsMsg'
Learn More

WWC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18337214 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337214 100 μg 100µL
18325286 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18337214 Supplier Novus Biologicals Supplier No. NBP315585100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WWC2 Polyclonal antibody specifically detects WWC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen WWC2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 50% glycerol, pH7.3
Gene Alias C2 and coiled-coil domain containing 2, FLJ34082, WW and C2 domain containing 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-430 of human WWC2 (NP_079225.5). DPDILKAEISTTRLRVKKLKRELSQMKQELLYKEQGFETLQQIDKKMSGGQSGYELSEAKAILTELKSIRKAISSGEKEKQDLMQSLAKLQERFHLDQNIGRSEPDLRCSPVNSHLCLSRQTLDAGSQTSISGDIGVRSRSNLAEKVRLSLQYEEAKRSMANLKIELSKLDSEAWPGALDIEKEKLMLINEKEELLKELQFVTPQKRTQDELERLEAERQRLEEELLSVRGTPSRALAERLRLEERRKELLQKLEETTKLTTYLHSQLKSL
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 80014
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.