missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WWC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545620
This item is not returnable.
View return policy
Description
Peptide sequence: SRELKPVGVM APASGPASTD AVSALLEQTA VELEKRQEGR SSTQTLEDSW Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 79%; Rat: 100%.
WWC1 (Protein KIBRA) is a probable regulator of the Hippo/SWH signaling pathway. This pathway plays a pivotal role in tumor suppression by restricting proliferation and promoting apoptosis. WWC1, along with NF2, can synergistically induce the phosphorylation of LATS1 and LATS2. It acts as a transcriptional coactivator of ESR1 - which plays an essential role in DYNLL1-mediated ESR1 transactivation. WWC1 also functions as a regulatior of collagen-stimulated activation of the ERK/MAPK cascade.
Specifications
| WWC1 | |
| Polyclonal | |
| Unconjugated | |
| WWC1 | |
| AA408228; AU017197; BC037006; FLJ10865; FLJ23369; HBeAg-binding protein 3; HBEBP3; HBEBP36; Kiaa0869; KIBRA; Kidney and brain protein; MEMRYQTL; mKIAA0869; PPP1R168; Protein KIBRA; protein phosphatase 1, regulatory subunit 168; protein WWC1; RGD1308329; WW and C2 domain containing 1; WW domain-containing protein 1; WW, C2 and coiled-coil domain containing 1; WWC1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23286 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IX03 | |
| WWC1 | |
| Synthetic peptide directed towards the C-terminal of human WWC1 (aa 793-842). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction