missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTAP Polyclonal Antibody

WTAP Polyclonal Antibody
Brand: Invitrogen PA581125
This item is not returnable.
View return policy
Description
WTAP Polyclonal Antibody for Western Blot, ELISA
Specifications
| WTAP | |
| Polyclonal | |
| Unconjugated | |
| WTAP | |
| 2810408K05Rik; 9430038B09Rik; Female-lethal(2)D homolog; hFL(2)D; KIAA0105; Mum2; PNAS-132; pre-mRNA-splicing regulator WTAP; putative pre-mRNA splicing regulator female-lethal(2D); RGD1563824; Wilms tumor 1 associated protein; wilms tumor 1-associating protein; Wilms' tumour 1-associating protein; Wilms tumour 1-associating protein; WT1-associated protein; WTAP | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 9589 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q15007 | |
| WTAP | |
| Recombinant Human WTAP protein (Met1-Arg151). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction