missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTAP Mouse anti-Human, Monkey, Clone:6B6B6, Abnova™
Mouse monoclonal antibody raised against recombinant human WTAP.
Brand: Abnova MAB16701.100ug
This item is not returnable.
View return policy
Description
The Wilms tumor suppressor gene WT1 appears to play a role in both transcriptional and posttranscriptional regulation of certain cellular genes. This gene encodes a WT1-associating protein, which is a ubiquitously expressed nuclear protein. Like WT1 protein, this protein is localized throughout the nucleoplasm as well as in speckles and partially colocalizes with splicing factors. Alternative splicing of this gene results in three transcript variants, two of which encode the same isoform. [provided by RefSeq]Specifications
| WTAP | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| WTAP | |
| Mouse | |
| 100 μg | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 6B6B6 | |
| Wilms tumor 1 associated protein | |
| In PBS (0.05% sodium azide) | |
| DKFZp686F20131, KIAA0105, MGC3925 | |
| Recombinant protein corresponding to amino acid 91-201 of human WTAP from E. coli. | |
| 9589 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction