missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTAP Antibody (OTI12G7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74870H
This item is not returnable.
View return policy
Description
WTAP Monoclonal specifically detects WTAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| WTAP | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| WTAP | |
| Recombinant protein fragment corresponding to amino acids 2-106 of human WTAP (NP_004897) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI12G7 | |
| Western Blot, Immunohistochemistry | |
| Female-lethal(2)D homolog, hFL(2)D, KIAA0105DKFZp686F20131, MGC3925, PNAS-132, pre-mRNA-splicing regulator WTAP, putative pre-mRNA splicing regulator female-lethal(2D), Wilms tumor 1 associated protein, Wilms tumor 1-associating protein, Wilms′ tumour 1-associating protein, WT1-associated protein | |
| Mouse | |
| Affinity Purified | |
| Cancer, Tumor Suppressors | |
| 9589.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction