missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Mouse anti-Human, Clone:5G11A5, Abnova™
Mouse monoclonal antibody raised against recombinant human WT1.
Brand: Abnova MAB16633.100ug
This item is not returnable.
View return policy
Description
This gene encodes a transcription factor that contains four zinc-finger motifs at the C-terminus and a proline/glutamine-rich DNA-binding domain at the N-terminus. It has an essential role in the normal development of the urogenital system, and it is mutated in a small subset of patients with Wilm′s tumors. Multiple transcript variants, resulting from alternative splicing at two coding exons, have been well characterized. There is also evidence for the use of non-AUG (CUG) translation initiation site upstream of, and in-frame with the first AUG, leading to additional isoforms. Authors of PMID:7926762 also provide evidence that WT1 mRNA undergoes RNA editing in human and rat, and that this process is tissue-restricted and developmentally regulated. [provided by RefSeq]Specifications
| WT1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| WT1 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 5G11A5 | |
| Wilms tumor 1 | |
| In PBS (0.05% sodium azide) | |
| GUD, WAGR, WIT-2, WT33 | |
| Recombinant protein corresponding to amino acid 314-479 of human WT1 from E. coli. | |
| 7490 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction