missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1, Mouse anti-Human, Clone: 1E9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007490-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: QDVRRVPGVAPTLVRSASETSEKRPFMCAYPGCNKRYFKLSHLQMHSRKHTGEKPYQCDFKDCERRFSRSDQLKRHQRRHTGVKPFQCKTCSpecifications
| WT1 | |
| Monoclonal | |
| Unconjugated | |
| Wilms tumor 1 | |
| GUD/WAGR/WIT-2/WT33 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1E9 | |
| In 1x PBS, pH 7.4 | |
| NM_000378 | |
| WT1 | |
| WT1 (NP_000369.3, 349 a.a. ∼ 439 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7490 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction