missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1-AS Polyclonal Antibody

WT1-AS Polyclonal Antibody
Brand: Invitrogen PA575590
This item is not returnable.
View return policy
Description
WT1-AS Polyclonal Antibody for Western Blot
Specifications
| WT1-AS | |
| Polyclonal | |
| Unconjugated | |
| WT1-AS | |
| 6030498F17; AI314831; Wilms tumor 1 homolog, opposite strand; WIT1; WIT-1; WT1 antisense RNA; WT1AS; Wt1-as; WT1-AS1; Wt1os | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 329480, 51352 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q06250 | |
| WT1-AS, Wt1os | |
| Synthetic peptide corresponding to residues in Human WT1-AS. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction