missing translation for 'onlineSavingsMsg'
Learn More

WT1 Antibody (SPM361) - IHC-Prediluted, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-48215

Product Code. 18154168

  • £375.00 / 7mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

WT1 Monoclonal specifically detects WT1 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WT1
Monoclonal
Unconjugated
1.0mM PBS and 0.05% BSA with 0.05% Sodium Azide
WT1
Recombinant protein corresponding to residues 1-181 of human WT1. (Uniprot: P19544)
7 mL
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors
7490
Human, Mouse
Purified
Immunohistochemistry (Paraffin)
SPM361
Immunohistochemistry-Paraffin
AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33
Mouse
Protein A or G purified
RUO
Primary
Recognizes a 47-55kDa-tumor suppressor protein, identified as Wilms tumor and mesothelioma. WT1 protein has been identified in proliferative mesothelial cells, malignant mesothelioma, ovarian carcinoma, gonadoblastoma, nephroblastoma, and desmoplastic small round cell tumor. Lung adenocarcinomas rarely stain positive with this antibody. WT1 protein expression in mesothelial cells has become a reliable marker for the diagnosis of mesotheliomas.
Store at 4C.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.