missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
WT1 Monoclonal antibody specifically detects WT1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
Specifications
| Antigen | WT1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Classification | Monoclonal |
| Clone | rWT1/857 |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human WT1 protein (Uniprot: P19544) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?