missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Antibody (CL13165), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18564-100ul
This item is not returnable.
View return policy
Description
WT1 Monoclonal antibody specifically detects WT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| WT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL13165 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
| This antibody was developed against a sythetic peptide for P19544. Actual immunogen sequence is proprietary. | |
| 100 μg | |
| Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
| 7490 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction