Learn More
WT1 Antibody (CL13165) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18564AF647
Product Code. 30294404
  • £599.00 / 0.10mL
Description
WT1 Monoclonal antibody specifically detects WT1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WT1
Monoclonal
Alexa Fluor 647
AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33
This antibody was developed using a synthetic peptide derived from P19544, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors
7490
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL13165
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified