missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Antibody (960522), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB5729
This item is not returnable.
View return policy
Description
WT1 Monoclonal specifically detects WT1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| WT1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 960522 | |
| Unconjugated | |
| PBS with No Preservative | |
| WT1 | |
| E. coli-derived recombinant human WT1 Met127-Gly249 Accession # P19544 | |
| 100 μg | |
| Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
| 7490 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction