missing translation for 'onlineSavingsMsg'
Learn More

WT-161, MedChemExpress

Product Code. 16495927
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495927 5 mg 5mg
16479746 50 mg 50mg
16474476 100 mg 100mg
16473198 10 mg 10mg
16472666 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495927 Supplier Medchem Express Supplier No. HY100871

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WT-161 is a potent and selective HDAC6 inhibitor with an IC50 of 0.40 nM. WT-161 also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).

WT-161 is a potent and selective HDAC6 inhibitor with an IC50 of 0.40 nM. WT-161 also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).

Specifications

Chemical Name or Material WT-161
Physical Form Solid
CAS 1206731-57-8
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C27H30N4O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (218.08 mM)
SMILES O=C(N/N=C/C1=CC=C(N(C2=CC=CC=C2)C3=CC=CC=C3)C=C1)CCCCCCC(NO)=O
Molecular Weight (g/mol) 458.55
Formula Weight 458.55
Percent Purity 98.52%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.