missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WSB1 Monoclonal Antibody (3E10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521610
This item is not returnable.
View return policy
Description
Peptide Sequence: QGHRTVKLVP WSQCLQNFLL HGTKNVTNSS SLRLPRQNSD GGQKNKPREH IIDCGDIVWS LAFGSSVPEK QSRCVNIEWH RFRFG.
WSB1 (WD repeat and SOCS box-containing 1), also known as SWIP1, is a 421 amino acid protein that contains one SOCS box domain and six WD repeats. Expressed as multiple alternatively spliced isoforms, WSB1 is thought to function as part of an SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex that mediates the ubiquitination and proteasomal degradation of target proteins, such as DIO2. Overexpression of WSB1 is implicated in pancreatic cancer progression, suggesting a role for WSB1 in carcinogenesis.
Specifications
| WSB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9Y6I7 | |
| WSB1 | |
| WSB1 (AAH21110.1, 53 a.a. ~ 137 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 3E10 | |
| Unconjugated | |
| WSB1 | |
| 1110056B13Rik; 2700038M07Rik; SOCS box-containing WD protein SWiP-1; SWIP1; WD repeat and SOCS box containing 1; WD repeat and SOCS box-containing 1; WD repeat and SOCS box-containing protein 1; WD repeat and SOCS box-containing protein 1-like protein; WD-40-repeat-containing protein with a SOCS box 1; WSB1; WSB-1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26118 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction