missing translation for 'onlineSavingsMsg'
Learn More

WS3, MedChemExpress

Product Code. 16485178
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485178 10 mg 10mg
16455876 100 mg 100mg
16401407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485178 Supplier Medchem Express Supplier No. HY12462

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WS3 is a novel proliferative molecule that promotes pancreatic β cell proliferation in rodent and human primary islets. WS3 can be used for the research of type 1 diabetes.

WS3 is a novel proliferative molecule that promotes pancreatic β cell proliferation in rodent and human primary islets. WS3 can be used for the research of type 1 diabetes.

Specifications

Chemical Name or Material WS3
CAS 1421227-52-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H30F3N7O3
Quantity 10 mg
Solubility Information DMSO : 2.2 mg/mL (3.86 mM; Need ultrasonic)
SMILES O=C(C1CC1)NC2=NC=NC(OC3=CC=C(NC(NC4=CC=C(CN5CCN(C)CC5)C(C(F)(F)F)=C4)=O)C=C3)=C2
Molecular Weight (g/mol) 569.58
Formula Weight 569.58
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.