missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WS 383
GREENER_CHOICE

Product Code. 18797056
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18797056

Brand: Tocris Bioscience™ 7055/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective and reversible DCN1-UBC12 interaction inhibitor

WS 383 is a potent and reversible DCN1-UBC12 interaction inhibitor (IC50 = 11 nM). The compound displays selectivity for DCN1-UBC12 over inhibition of a panel of kinases. including Bruton's tyrosine kinase (BTK), cyclin-dependent kinases (CDKs) and epidermal growth factor receptor (EGFR). In vitro, WS 383 selectively inhibits cullin-3 neddylation over other cullins, and induces accumulation of p21, p27 and NRF2.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[[2-[[(4-Chlorophenyl)methyl]thio]-5-methyl[1,2,4 ]triazolo[1,5-a]pyrimidin-7-yl]thio]-N,N-dimethyl-1 H-tetrazole-1-ethanamine hydrochloride
CAS 2247544-02-9
Quantity 50 mg
Target Ubiquitin Ligase (E3) Inhibitors
Molecular Formula C18H20ClN9S2.HCl
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.