missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WS 12

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent TRPM8 agonist that acts as a cooling agent (EC50 = 193 nM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 289.41 |
| Purity | >99% |
| Molecular Weight (g/mol) | 289.419 |
| Chemical Name or Material | WS 12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction