missing translation for 'onlineSavingsMsg'
Learn More

WS-12, MedChemExpress

Product Code. 16454198
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454198 10 mg 10mg
16426917 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16454198 Supplier Medchem Express Supplier No. HY108449
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress WS-12 (AR-15512) is an agonist of TRPM8 with an EC50 of 39 nM.

WS-12 (AR-15512) is an agonist of TRPM8 with an EC50 of 39 nM.

Specifications

Chemical Name or Material WS-12
CAS 68489-09-8
Color White
Purity Grade Notes Research
Molecular Formula C18H27NO2
Quantity 10 mg
Synonym AR-15512 AVX-012
Solubility Information DMSO : 20 mg/mL (69.11 mM; Need ultrasonic)
SMILES O=C([C@H]1[C@H](C(C)C)CC[C@@H](C)C1)NC2=CC=C(OC)C=C2
Molecular Weight (g/mol) 289.41
Formula Weight 289.41
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.