missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WRN Recombinant Rabbit Monoclonal Antibody (JE52-72)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536149
This item is not returnable.
View return policy
Description
WRN Recombinant Monoclonal Antibody for Western Blot
This gene encodes a member of the RecQ subfamily and the DEAH (Asp-Glu-Ala-His) subfamily of DNA and RNA helicases. DNA helicases are involved in many aspects of DNA metabolism, including transcription, replication, recombination, and repair. This protein contains a nuclear localization signal in the C-terminus and shows a predominant nucleolar localization. It possesses an intrinsic 3' to 5' DNA helicase activity, and is also a 3' to 5' exonuclease. Based on interactions between this protein and Ku70/80 heterodimer in DNA end processing, this protein may be involved in the repair of double strand DNA breaks. Defects in this gene are the cause of Werner syndrome, an autosomal recessive disorder characterized by premature aging.
Specifications
| WRN | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q14191 | |
| WRN | |
| Recombinant protein within Human WRN aa 1,260-1,432. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| JE52-72 | |
| Unconjugated | |
| WRN | |
| AI846146; DKFZp686C2056; DNA helicase, RecQ-like type 3; exonuclease WRN; RecQ protein-like 2; RecQ3; RECQL2; RECQL3; RGD1564788; similar to Werner syndrome helicase homolog; Werner syndrome ATP-dependent helicase; Werner syndrome ATP-dependent helicase homolog; Werner syndrome homolog (human); Werner syndrome RecQ like helicase; Werner syndrome, RecQ helicase-like; WRN | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7486 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction