missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WRN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20914
This item is not returnable.
View return policy
Description
WRN Polyclonal specifically detects WRN in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| WRN | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
| DKFZp686C2056, DNA helicase, RecQ-like type 3, EC 3.1, EC 3.6.1, EC 3.6.4.12, Exonuclease WRN, RecQ protein-like 2, RecQ3, RECQ3RECQL3, RECQL2Werner syndrome ATP-dependent helicase, Werner syndrome, Werner syndrome, RecQ helicase-like | |
| Rabbit | |
| 162 kDa | |
| 0.1 mL | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Tumor Suppressors | |
| 7486 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500 - 1:3000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000 | |
| Q14191 | |
| WRN | |
| Recombinant protein encompassing a sequence within the C-terminus region of human WRN. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction