missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WRAP53 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106452
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total WRAP53.
TXNDC1 is a thioredoxin -related protein with disulfide reductase activity.
Specifications
| WRAP53 | |
| Polyclonal | |
| Unconjugated | |
| WRAP53 | |
| BC021790; DKCB3; TCAB1; Telomerase Cajal body protein 1; WD repeat containing antisense to TP53; WD repeat containing, antisense to TP53; WD repeat containing, antisense to Trp53; WD repeat domain 79; WD repeat-containing protein 79; WD repeat-containing protein WRAP53; WD40 protein Wrap53; WD40 repeat-containing protein antisense to TP53; WD40 repeat-containing protein antisense to TP53 gene; WD40 repeat-containing protein antisense to TP53 gene homolog; WD40 repeat-containing protein encoded by RNA antisense to p53; WD40 repeat-containing protein encoding RNA antisense to p53; Wdr79; WRAP53; WRAP53beta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216853, 287432, 55135 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5XII5, Q8VC51, Q9BUR4 | |
| WRAP53 | |
| A synthesized peptide derived from human WRAP53(Accession Q9BUR4), corresponding to amino acid residues F150-P200. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction