missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WR 1065
GREENER_CHOICE

Product Code. 18004872
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18004872

Brand: Tocris Bioscience™ 3356/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p53 activator. Also ROS scavenger

Cell-permeable reactive oxygen species scavenger that activates p53 through a JNK-dependent signaling pathway. Activates p21waf-1 and MDM2, represses transcription of Myc and thymidine kinase, and inhibits DNA topoisomerase IIα. Induces cell cycle arrest and exhibits cytoprotective activity towards normal cells but not cancer cells in vivo. Also exhibits broad spectrum antiviral activity in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14653-77-1
Molecular Formula C5H16Cl2N2S

Specifications

Quantity 10 mg
Formula Weight 207.16
Purity >98%
Molecular Weight (g/mol) 207.16
Chemical Name or Material WR 1065
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.