missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WQ 1
Description
WQ 1 is a high affinity and selective σ1 receptor antagonist (pKi values are 10.28-10.85, 5.81-6.53 and 5.80 at σ1, σ2 and DAT). Displays σ1/σ2 selectivity ratio of 29512. Selectively blocks binge eating without a general inhibition of food intake in rats.
Specifications
Specifications
| Chemical Name or Material | 2,3-Dihydro-2-[1-(phenylmethyl)-4-piperidinyl]-4 H-1-benzothiopyran-4-one |
| CAS | 1204401-49-9 |
| Quantity | 10 mg |
| Target | Sigma1 Receptor Antagonists |
| Molecular Formula | C21H23NOS |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?