missing translation for 'onlineSavingsMsg'
Learn More

Manutan Woven polyester strapping. Wxl 16mm x 850m

Brand:  Manutan A329094

Encompass_Preferred

Woven polyester strapping. Wxl 16mm x 850m

Product Code. 12802219

  • £72.60 / Pack of 1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.