missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Wortmannin

Product Code. 17160242
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17160242 1 mg 1mg
17170242 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17160242 Supplier STEMCELL Technologies Supplier No. 73562
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

PI3K/AKT pathway inhibitor; Inhibits PI3K and PLKs

  • Wortmannin is a fungal metabolite that covalently binds to and inhibits phosphatidylinositol-3-kinases (PI3K) of class I, II, and III
  • Species-specific differences in the class II PI3Ks determine sensitivity with IC50 = 5, 50, and 450 nM for Drosophila, mouse, and human, respectively (Fruman et al.; Wymann et al.; Okada et al.) Wortmannin also inhibits polo-like kinases (PLK) PLK1 and PLK3 with IC50 = 24 and 49 nM, respectively (Liu et al. 2005; Liu et al. 2007)
  • At high concentrations it can also inhibit other kinases such as mammalian target of rapamycin (mTOR), DNA-dependent protein kinase catalytic subunit (DNA-PKcs), phosphatidylinositol-4-kinase (PI4K), myosin light-chain kinase (MLCK), and mitogen-activated protein kinase (MAPK; Fruman et al.; Meyers & Cantley; Hartley et al.; Brunn et al.; Nakanishi et al.)

Specifications

CAS 19545-26-7
Synonym KY 12420
Quantity 1 mg
Molecular Formula C23H24O8
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.