missing translation for 'onlineSavingsMsg'
Learn More

Techsit Worktech PU Laboratory Chair

Product Code. 30306787
Encompass_Preferred
Change view
Click to view available options
Seating Type:
Desk Height
High Bench Height
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Seating Type Unit Size
30306787 Desk Height Each
30306775 High Bench Height Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30306787 Supplier Techsit Supplier No. TS1420
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PU Lab chair

  • PU high chair with adjustable foot ring on glides
  • Hard wearing, easy-to-clean PU seat and back
  • Fully adjustable seat height, back height, and back rake
  • Ergonomically designed and contoured seat and back
  • Cleanroom
  • Class 5 suitable

PU, Lab Chair

Specifications

Product Type Lab Swivel Chair
Color Black
Autoclavable Not Autoclavable
Back Style Ergonomic
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support Yes
With Tilt No
Material Polyurethane
Seating Type Desk Height
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.