missing translation for 'onlineSavingsMsg'
Learn More

Techsit Worktech Laboratory Vinyl Low Chair

Product Code. 30306777
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Blue
Style:
High back
On castors
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Style unitSize
30306777 Black High back Each
30306776 Black On castors Each
30306786 Blue On castors Each
30306782 Blue High back Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30306777 Supplier Techsit Supplier No. TS1320HB/BLK

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Low antimicrobial vinyl Lab chair

  • Vinyl
  • Laboratory low chair on castors
  • Antimicrobial Vinyl that is easy to clean
  • Fully adjustable seat height, back height, and back rake
  • Ergonomically designed and contoured seat and back
  • Cleanroom
  • Class 7 suitable

Specifications

Product Type Lab Swivel Chair
Color Black
Autoclavable Not Autoclavable
Back Style Ergonomic
Style High back
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support Yes
With Tilt No
Material Vinyl Upholstery
Seating Type Desk Height
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.