missing translation for 'onlineSavingsMsg'
Learn More

Techsit Worktech Laboratory Vinyl High Chair

Product Code. 30306788
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Blue
Style:
Footrings on glides
High back
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Style Unit Size
30306788 Blue High back Each
30306783 Black Footrings on glides Each
30306778 Blue Footrings on glides Each
30306780 Black High back Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30306788 Supplier Techsit Supplier No. TS1311HB/Blue
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High antimicrobial vinyl Lab chair

  • Vinyl
  • Laboratory high chair with adjustable foot ring on glides
  • Antimicrobial Vinyl that is easy to clean
  • Fully adjustable seat height, back height, and back rake
  • Ergonomically designed and contoured seat and back
  • Cleanroom
  • Class 7 suitable

Specifications

Product Type Lab Swivel Chair
Color Blue
Autoclavable Not Autoclavable
Back Style Ergonomic
Style High back
With Armrest No
With Backrest Yes
With Casters No
Footrest Yes
With Lumbar Support Yes
With Tilt No
Material Vinyl Upholstery
Seating Type High Bench Height
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.