missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Workstation Custom Designer Software

Product Code. 15805737
Click to view available options
:
Custom Designer Software; Operating System: Windows™ 10, Windows™ 7, Windows™ 8; For Use With (Application): Barcode and Electrical Labeling, General Identification, Inventory Labeling
This item is not returnable. View return policy

Product Code. 15805737

Brand: Brady™ 149195

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Software for the custom design and development of professional labels.

When you need full label creation capabilities to streamline your work and maximize your investment, Custom Designer is the app of choice. This premier label editing app includes all of the features of Custom Designer Lite, plus more barcodes, linked objects, label error checking, subscript and superscript features, advanced editing tools and more.

  • Opens saved 'graphics mode' files from LabelMark 5 and 6 without the need to convert
  • Compatible with: BBP™11 Label Printer, BBP™12 Industrial Label Printer, BBP™30 Sign & Label Printer, BBP™31 Sign & Label Printer, BBP™33 Sign & Label Printer, BBP™35 Multicolor Label Printer, BBP™37 Multicolor & Cut Label Printer, BBP™85 Sign & Label Printer, BMP™41 Label Printer, BMP™51 Label Printer, BMP™53 Label Printer, BMP™61 Label Printer, BMP™71 Label Printer, BradyJet J2000 Color Label Printer, BradyJet J4000 Color Label Printer, BradyJet J5000 Color Label Printer, BradyPrinter i3300 Industrial Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, BradyPrinter M611 Mobile Label Printer, BradyPrinter PR Plus Series Label Printer, BradyPrinter S3000 Sign & Label Printer, GlobalMark™ 2 Multicolor Industrial Label Maker, IP™ Series Printer, THT 76 mm Core Printers, Wraptor™ A6500 Wrap Printer Applicator, Wraptor™ Wire Marking System

Specifications

Format Email with Activation Code
Languages Bulgarian, Chinese (Tradditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Norwegian, Polish, Portuguese, Romanian, Russian, Slovakian, Slovenian, Spanish, Swedish, Turkish
Software Type Sign and Label Design Software
Operating System Windows™ 10, Windows™ 7, Windows™ 8
For Use With (Application) Barcode and Electrical Labeling, General Identification, Inventory Labeling
Alternate Part Number BWS-CUD-EM
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.