missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Works Calibration Certificate - Factory 160°C

Factory calibration certificate +160°C

Brand:  Memmert™ WTB24D00123

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 17120365

  • £279.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products

Description

Description

  • Accessory
  • Factory calibration certificate
  • 160°C
Specifications

Specifications

WTB24
Works Calibration Certificate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers